missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Straight-sided Beaker Set

Ideal for general laboratory use - straight stable shape

Brand:  Azlon BDA218MES

 View more versions of this product

Product Code. 10580681

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Ideal for general laboratory use - straight stable shape

  • Set of popular classic Azlon beakers
  • Polypropylene material - good chemical resistance
  • Can withstand temperatures up to 121°C and therefore autoclavable
  • Graduations are clearly printed in indelible solvent resistant ink
TRUSTED_SUSTAINABILITY
Specifications

Specifications

50 mL, 100 mL, and 250 mL
Ideal for General Laboratory Use
Straight Sided
Yes
Polypropylene
Set of 3 Beakers, (50, 100 and 250 mL)
Autoclavable
ISO 7056
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.