Learn More
Azlon™ Straight-sided Beaker Set
Ideal for general laboratory use - straight stable shape
7.33 € - 38.15 €
Specifications
Material Polypropylene
For Use With (Application) Ideal for General Laboratory Use
Type Straight Sided
Autoclavable Autoclavable
Graduated Yes
View More Specs
Products 6
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10580681
View Documents
Azlon
BDA218MES
50 mL, 100 mL, and 250 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12483551
View Documents
Azlon
BDA218MES
50 mL, 100 mL, and 250 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12473551
View Documents
Azlon
BDA1198
500 mL, 1000 mL, and 2000 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10578570
View Documents
Azlon
BDA1199
10 mL, 25 mL, 50 mL, 100 mL, and 250 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10293430
View Documents
Azlon
BDA1200
10 mL, 25 mL, 50 mL, 100 mL, 250 mL, 500 mL, 1000 mL, and 2000 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10165824
View Documents
Azlon
BDA300P
10 mL, 25 mL, 50 mL, 100 mL, 250 mL, 400 mL, 500 mL, 600 mL, 1000 mL, and 2000 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Ideal for general laboratory use - straight stable shape
  • Set of popular classic Azlon beakers
  • Polypropylene material - good chemical resistance
  • Can withstand temperatures up to 121°C and therefore autoclavable
  • Graduations are clearly printed in indelible solvent resistant ink
Specifications
Specifications
Polypropylene
Straight Sided
Yes
Ideal for General Laboratory Use
Autoclavable
ISO 7056