missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ B-7610 Paper Hazardous Substance Labels, 25 mm Core
Pre-printed, desktop printer labels for CLP-GHS hazardous substances.
Brand: Brady™ 622318
This item is not returnable.
View return policy
Description
- Supplied with pre-printed red diamonds
- Ready for you to print the required warning, compliant to the latest regulations
Specifications
Not Autoclavable | |
Permanent Acrylic | |
Matte | |
Thermal Transfer | |
Hazard Communication Identification | |
1 | |
Rectangle | |
0.081 mm | |
55 mm |
THTCLP-S-L4B-7610-0.4-SC | |
Red on White | |
Paper (B-7610) | |
Non-sterile | |
BBP™11, BBP™12, and BradyPrinter PR Plus Series Label Printers, THT 76 mm Core Printers | |
400 Labels | |
-40°C to +50°C | |
91 mm | |
85 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Brady™ B-7610 Paper Hazardous Substance Labels, 25 mm Core > Paper (B-7610) Hazardous Substance Labels, 25 mm Core; Color: Red on White; Finish: Matte; Height (Metric): 85 mm; Vertical Repeat (Metric): 91 mm; Width (Metric): 55 mm
Spot an opportunity for improvement?Share a Content Correction