missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ B-7610 Paper Hazardous Substance Labels, 25 mm Core
Pre-printed, desktop printer labels for CLP-GHS hazardous substances.
129.00 € - 136.00 €
Specifications
Autoclavable | Not Autoclavable |
---|---|
Adhesive Type | Permanent Acrylic |
Color | Red on White |
Finish | Matte |
Material | Paper (B-7610) |
Product Code | Brand | Height (Metric) | Vertical Repeat (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Vertical Repeat (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
16637763
|
Brady™
622318 |
85 mm | 91 mm | 55 mm |
129.00 €
1 roll |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||
16647763
|
Brady™
622330 |
110 mm | 115.4 mm | 75 mm |
136.00 €
1 roll |
In Stock
Log in to see stock. |
||||
Description
- Supplied with pre-printed red diamonds
- Ready for you to print the required warning, compliant to the latest regulations
Specifications
Not Autoclavable | |
Red on White | |
Paper (B-7610) | |
Non-sterile | |
BBP™11, BBP™12, and BradyPrinter PR Plus Series Label Printers, THT 76 mm Core Printers | |
Rectangle | |
0.081 mm |
Permanent Acrylic | |
Matte | |
Thermal Transfer | |
Hazard Communication Identification | |
1 | |
-40°C to +50°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Brady™ B-7610 Paper Hazardous Substance Labels, 25 mm Core