Learn More
CPC™ Back-to-Back Body Adapter
MPC Body x MPX Back-to-Back Body Adapter
Brand:  CPC™ MPC17X1739
Product Code. 17425103
  • 1350.00 € / Pack of 25
Estimated Shipment: 30-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Medical Grade
  • Non-valved
  • Natural finish
Specifications
Specifications
USP Class VI, ADCF
Polysulfone
Gamma, Autoclave
-40°F
Amber
Up to 60 psi, 4.1 bar
300°F
149°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
CPC™ Back-to-Back Body Adapter > Polysulfone, USP Class VI, ADCF, sterlizations: Gamma Autoclave, free floating, -40 F to 300 F