missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Microplate Rack

Save valuable lab space with stackable racks for tubes and plates

Brand:  Eppendorf™ M1289-0700

Product Code. 11349864

  • 382.52 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Compatible with 0.1mL, 0.2mL, 0.5mL tubes and 96-well plates
  • Can be stored in temperatures as low as -90°C
  • Autoclavable (121°C, 20 min.)
  • Stackable
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Not Autoclavable
Plates
3 Deep Well and 9 Standard
Biological Shakers
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Eppendorf™ Microplate Rack > Microplate Rack; Stackable; 3 dwp/ 9 standard plates

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.