missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Bacto™ Casitone
An animal origin (AO) pancreatic enzymatic digest of casein.
197.00 €
Specifications
Content And Storage | Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. |
---|---|
Form | Powder |
Product Type | Dehydrated Culture Media |
For Use With (Application) | Production of Escherichia coli-based recombinant proteins |
Product Line | Bacto™ |
Description
- An animal origin (AO) pancreatic enzymatic digest of casein similar to Difco Casein Digest
- This supplement provides a nutritional profile based on casein
- The specific enzymatic digest performed for this product results in a particularly high content of amino acids and peptides of varying sizes, making it a more nutritious hydrolysate
- It has been successfully used for the production of Escherichia coli-based recombinant proteins
- Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
Specifications
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. | |
Dehydrated Culture Media | |
Bacto™ |
Powder | |
Production of Escherichia coli-based recombinant proteins |
Safety and Handling
missing translation for 'shelfLife' : 1825 Days