Learn More
Tocris Bioscience™ BAG 956
Dual PI 3-kinase and PDPK1 (PDK1) inhibitor
Brand:  Tocris Bioscience™ 3606/10
 View more versions of this product
Product Code. 18025612
  • 240.00 € / 10mg
Explore more special offers
Description
Description
BAG 956 is a dual PDPK1 (PDK1) and class I PI 3-K inhibitor (IC50 values are 245, 56, 446, 35 and 117 nM for PDPK1 and PI 3-K p110 -α, -β, -δ, and -γ respectively). Inhibits cellular AKT phosphorylation at Thr308. Blocks cell proliferation, causing arrest in G1 phase of the cell cycle. Slows tumor progression in mouse xenograft models.
Specifications
Specifications
98%
10 mg
427.5
Store at -20°C
BAG 956
BAG 956