Learn More
Sartorius Balance Printers
Transfer data automatically or manually. Sartorius Balance Printers record individual weighing data and generate a statistical evaluation of weighing data. Values transferred from an on-line weighing instrument are stored.
Brand: Sartorius YDP20-0CE
Description
Transfer data automatically or manually. Sartorius™ Balance Printers record individual weighing data and generate a statistical evaluation of weighing data. Values transferred from an on-line weighing instrument are stored.
- Built-in real-time clock
- Built-in statistics program
- Counter for consecutive numbering of transactions can be activated
- High contrast LCD shows date or time as well as current transaction number
- Integrated rechargeable batteries with battery level indicator
- Ultrafast print head
- Runs on the balance's power; no separate power supply necessary
- Easy to change paper roll and ink ribbon cassette
Specifications
CPA Balance, Extended Balance, Moisture Analyzers | |
215 mm | |
78 mm | |
0.9 kg |
215 x 155 x 78 mm | |
155 cm | |
interface cable, Paper roll, four rechargeable b | |
Data Printer |