Learn More
Sartorius Balance Printers
Transfer data automatically or manually. Sartorius Balance Printers record individual weighing data and generate a statistical evaluation of weighing data. Values transferred from an on-line weighing instrument are stored.
938.00 € - 1210.00 €
Product Code | Brand | For Use With (Equipment) | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
10239151
![]() |
Sartorius
YDP20-0CE |
CPA Balance, Extended Balance, Moisture Analyzers | 215 x 155 x 78 mm |
1210.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10506089
![]() |
Sartorius
YDP10-0CE |
CPA Balance, Extended Balance, Moisture Analyzers. (For use w/ Cubis 1 MSE or MSA models.) | 230 x 158 x 80 mm |
938.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Transfer data automatically or manually. Sartorius™ Balance Printers record individual weighing data and generate a statistical evaluation of weighing data. Values transferred from an on-line weighing instrument are stored.
- Built-in real-time clock
- Built-in statistics program
- Counter for consecutive numbering of transactions can be activated
- High contrast LCD shows date or time as well as current transaction number
- Integrated rechargeable batteries with battery level indicator
- Ultrafast print head
- Runs on the balance's power; no separate power supply necessary
- Easy to change paper roll and ink ribbon cassette
Specifications
Data Printer |