Learn More
Corning™ Bar-coded Cryogenic Vials
For storing biological and compound samples in labeled vials at cryogenic temperatures. Corning™ Bar-coded Cryogenic Vials provides a validated unique bar-code that will not fade or fall off.
Brand:  Corning™ 8672
 View more versions of this product
Product Code. 15515729
  • 382.28 € / Pack of 500
Estimated Shipment: 09-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from polypropylene
  • Leak proof seal for integrity of storage
  • Autoclavable vials and caps, can be re-sterilized
  • Unique 1D, 2D bar-codes and human readable characters
  • Features star shaped cavity in cap and is compatible with decapping equipment
  • Locking base stabilizes tubes for automated handling
  • Graduations and writing patch for manual use also
  • Each tube is checked for clarity and readability of 2D code
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Orange
Sterile
Cell biology, cell banking, compound cryopreservation
50
Round, Self-standing
Polyethylene
Natural
2 mL
500
Internal
1D
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Bar-coded Cryogenic Vials > Internal; 1D