missing translation for 'onlineSavingsMsg'
Learn More

Barium, 99+%, rods, ∽2cm diameter, Thermo Scientific Chemicals

199.00 € - 1044.00 €

Chemical Identifiers

CAS 7440-39-3
Molecular Formula Ba
Molecular Weight (g/mol) 137.33
MDL Number MFCD00134031
InChI Key DSAJWYNOEDNPEQ-UHFFFAOYSA-N
Synonym bario, baryum, bario spanish, baryum french, barium, soluble compounds, unii-24gp945v5t, and soluble compounds, barium, elemental, and compounds, atom
PubChem CID 5355457
ChEBI CHEBI:32595
IUPAC Name barium
SMILES [Ba]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10712973
View Documents Promotion Details
Thermo Scientific Acros
317861000
100 g
199.00 €
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10331484
View Documents Promotion Details
Thermo Scientific Acros
317865000
500 g
1044.00 €
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-39-3
137.33
DSAJWYNOEDNPEQ-UHFFFAOYSA-N
5355457
barium
Ba
MFCD00134031
bario, baryum, bario spanish, baryum french, barium, soluble compounds, unii-24gp945v5t, and soluble compounds, barium, elemental, and compounds, atom
CHEBI:32595
[Ba]

Specifications

Gray
725.0°C
99.0
99+%
Ba
15, 960
DSAJWYNOEDNPEQ-UHFFFAOYSA-N
barium
5355457
137.34
Plastic bottle
Barium, 99+%
7440-39-3
1640.0°C
100.0
Ba
MFCD00134031
bario, baryum, bario spanish, baryum french, barium, soluble compounds, unii-24gp945v5t, and soluble compounds, barium, elemental, and compounds, atom
[Ba]
137.33
CHEBI:32595
≥99%
Rod
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable solid.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
In contact with water releases flammable gas.
Reacts violently with water.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

Product Identifier
  • Barium in Paraffin oil

Signal Word
  • Danger

Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 1
  • Aspiration toxicity Category 1

Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H304-May be fatal if swallowed and enters airways.

Precautionary Statement
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

Supplemental information
  • MIXTURE LIST-Contains : White mineral oil, Barium

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only