Learn More
Barium acetate, ACS, 99.0-102.0%, Thermo Scientific Chemicals
59.80 € - 544.71 €
Chemical Identifiers
CAS | 543-80-6 |
---|---|
Molecular Formula | C4H6BaO4 |
Molecular Weight (g/mol) | 255.42 |
MDL Number | MFCD00012447 |
InChI Key | ITHZDDVSAWDQPZ-UHFFFAOYSA-L |
Synonym | barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate |
PubChem CID | 10980 |
IUPAC Name | barium(2+);diacetate |
SMILES | [Ba++].CC([O-])=O.CC([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11366598
|
Thermo Scientific Alfa Aesar
012198.30 |
250 g |
59.80 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11346598
|
Thermo Scientific Alfa Aesar
012198.A1 |
1 kg |
132.66 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11356598
|
Thermo Scientific Alfa Aesar
012198.A7 |
5 kg |
544.71 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Barium acetate is used as a chemical reagent, textile mordant and lubricant. It is used to prepare other acetates and barium carbonate. It acts as a mordant for printing textile fabrics. It is utilized for drying paints and varnishes. It serves as a catalyst in organic synthesis. It plays an important role in the separation of glycosaminoglycans. Further, it acts as a cationization agent for sensitive multiple reaction monitoring of plasma free fatty acid profiling in a fish oil human intervention. In addition, it is used in the process of titrating sulfates of quinine and quinidine in nonaqueous conditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
543-80-6 | |
255.42 | |
ITHZDDVSAWDQPZ-UHFFFAOYSA-L | |
10980 | |
[Ba++].CC([O-])=O.CC([O-])=O |
C4H6BaO4 | |
MFCD00012447 | |
barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate | |
barium(2+);diacetate |
Specifications
543-80-6 | |
C4H6BaO4 | |
UN1564 | |
barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate | |
ITHZDDVSAWDQPZ-UHFFFAOYSA-L | |
barium(2+);diacetate | |
10980 | |
99.0 to 102.0% | |
Odorless | |
Barium acetate |
Crystalline | |
MFCD00012447 | |
14,965 | |
Soluble in water. Insoluble in alcohol. | |
[Ba++].CC([O-])=O.CC([O-])=O | |
255.42 | |
255.43 | |
ACS | |
2.47 g/mL |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
missing translation for 'einecsNumber' : 208-849-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only