Learn More
Barium chloride dihydrate, ACS, Thermo Scientific Chemicals
37.49 € - 112.56 €
Chemical Identifiers
CAS | 10326-27-9 |
---|---|
Molecular Formula | BaCl2H4O2 |
Molecular Weight (g/mol) | 244.26 |
MDL Number | MFCD00149154 |
InChI Key | PWHCIQQGOQTFAE-UHFFFAOYSA-L |
Synonym | barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d |
PubChem CID | 5284346 |
ChEBI | CHEBI:86153 |
SMILES | O.O.[Cl-].[Cl-].[Ba++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11371669
|
Thermo Scientific Alfa Aesar
012310.36 |
500 g |
37.49 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11381669
|
Thermo Scientific Alfa Aesar
012310.A3 |
2 kg |
112.56 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Barium chloride is usually employed as a chemical reagent to precipitate soluble sulfates and also in the purification of the seawater solution. It is practiced in several applications like heat-treating baths, waste water treatment, the production of PVC stabilizers, oil lubricants, fireworks materials, hardening of steel, in the manufacture of pigments and in the manufacture of other barium salts. It is applied to prepare composites to absorb ammonia and to increase the crystallinty of polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10326-27-9 | |
244.26 | |
PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
5284346 | |
O.O.[Cl-].[Cl-].[Ba++] |
BaCl2H4O2 | |
MFCD00149154 | |
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
CHEBI:86153 |
Specifications
113°C-2H2O | |
Crystalline | |
MFCD00149154 | |
14,971 | |
Very soluble in water; Soluble in methanol; Insoluble in ethanol,acetone,ethyl acetate. | |
O.O.[Cl-].[Cl-].[Ba++] | |
244.26 | |
CHEBI:86153 | |
ACS | |
Barium chloride, dihydrate |
10326-27-9 | |
BaCl2H4O2 | |
UN1564 | |
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
barium(2+) dihydrate dichloride | |
5284346 | |
244.28 (208.25 Anhydrous) | |
3.097 |
Safety and Handling
GHS H Statement
H301-H332
P261-P264b-P270-P271-P280i-P301+P310-P304+P340-P305+P351+P338-P312-P330-P501c
H301-H319-H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
missing translation for 'einecsNumber' : 233-788-1
missing translation for 'rtecsNumber' : CQ8751000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Barium chloride dihydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Barium chloride dihydrate
RUO – Research Use Only