Learn More
Barium chloride, ultra dry, 99.95% (metals basis), Thermo Scientific™
82.00 € - 338.00 €
Chemical Identifiers
CAS | 10361-37-2 |
---|---|
Molecular Formula | BaCl2 |
Molecular Weight (g/mol) | 208.23 |
MDL Number | MFCD00003445 |
InChI Key | WDIHJSXYQDMJHN-UHFFFAOYSA-L |
Synonym | barium chloride, barium dichloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508 |
PubChem CID | 25204 |
ChEBI | CHEBI:63317 |
SMILES | [Cl-].[Cl-].[Ba++] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Ampouled under argon
Chemical Identifiers
10361-37-2 | |
208.23 | |
WDIHJSXYQDMJHN-UHFFFAOYSA-L | |
25204 | |
[Cl-].[Cl-].[Ba++] |
BaCl2 | |
MFCD00003445 | |
barium chloride, barium dichloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508 | |
CHEBI:63317 |
Specifications
1560°C | |
10361-37-2 | |
Beads | |
MFCD00003445 | |
14,971 | |
Soluble in water 370g/L. | |
[Cl-].[Cl-].[Ba++] | |
208.23 | |
CHEBI:63317 | |
99.95% | |
Ampule | |
3.9 g/mL | |
Barium chloride, Ultra dry |
963°C | |
White | |
BaCl2 | |
UN1564 | |
barium chloride, barium dichloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508 | |
WDIHJSXYQDMJHN-UHFFFAOYSA-L | |
barium(2+) dichloride | |
25204 | |
208.25 | |
Odorless | |
Hygroscopic | |
(metals basis) |
Safety and Handling
GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P310-P304+P340-P312-P330-P501c
H301-H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
missing translation for 'einecsNumber' : 233-788-1
missing translation for 'rtecsNumber' : CQ8750000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Ampouled under argon
Recommended Storage : Ambient temperatures; Store under Argon
Your input is important to us. Please complete this form to provide feedback related to the content on this product.