Learn More
Barium iodide hydrate, 95%, Thermo Scientific Chemicals
Light Sensitive and Hygroscopic
75.48 € - 245.22 €
Chemical Identifiers
CAS | 13718-50-8 |
---|---|
Molecular Formula | BaI2 |
Molecular Weight (g/mol) | 391.14 |
MDL Number | MFCD00003452 |
InChI Key | SGUXGJPBTNFBAD-UHFFFAOYSA-L |
IUPAC Name | barium(2+) diiodide |
SMILES | [I-].[I-].[Ba++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11387098
|
Thermo Scientific Alfa Aesar
015126.14 |
25 g |
75.48 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11377098
|
Thermo Scientific Alfa Aesar
015126.22 |
100 g |
245.22 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Useful in pyrotechnic devices. A reagent for facile synthesis of poly(phenylcarbyne).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13718-50-8 | |
391.14 | |
SGUXGJPBTNFBAD-UHFFFAOYSA-L | |
[I-].[I-].[Ba++] |
BaI2 | |
MFCD00003452 | |
barium(2+) diiodide |
Specifications
13718-50-8 | |
Powder | |
BaI2 | |
Freely soluble in water. Soluble in alcohol and acetone | |
[I-].[I-].[Ba++] | |
391.14 | |
Odorless | |
Barium iodide hydrate |
White | |
95% | |
MFCD00003452 | |
SGUXGJPBTNFBAD-UHFFFAOYSA-L | |
barium(2+) diiodide | |
391.15 (Anhydrous) | |
Hygroscopic; Light sensitive |
Safety and Handling
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III
missing translation for 'einecsNumber' : 237-276-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.