Learn More
Barium isopropoxide, Thermo Scientific Chemicals
41.00 € - 496.47 €
Chemical Identifiers
CAS | 24363-37-9 |
---|---|
Molecular Formula | C6H14BaO2 |
Molecular Weight (g/mol) | 255.503 |
MDL Number | MFCD00050486 |
InChI Key | CPUJSIVIXCTVEI-UHFFFAOYSA-N |
Synonym | barium isopropoxide, barium 2+ ; propan-2-olate, barium diisopropoxide, acmc-20alr0, barium ii isopropoxide, barium 2+ bis propan-2-olate, barium 2+ ion bis propan-2-olate |
PubChem CID | 6101105 |
IUPAC Name | barium(2+);propan-2-olate |
SMILES | CC(C)[O-].CC(C)[O-].[Ba+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11352478
|
Thermo Scientific Alfa Aesar
014523.03 |
1 g | N/A | ||||||
11332478
|
Thermo Scientific Alfa Aesar
014523.06 |
5 g | N/A | ||||||
11342478
|
Thermo Scientific Alfa Aesar
014523.14 |
25 g |
496.47 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
24363-37-9 | |
255.503 | |
CPUJSIVIXCTVEI-UHFFFAOYSA-N | |
6101105 | |
CC(C)[O-].CC(C)[O-].[Ba+2] |
C6H14BaO2 | |
MFCD00050486 | |
barium isopropoxide, barium 2+ ; propan-2-olate, barium diisopropoxide, acmc-20alr0, barium ii isopropoxide, barium 2+ bis propan-2-olate, barium 2+ ion bis propan-2-olate | |
barium(2+);propan-2-olate |
Specifications
∼200°C (decomposition) | |
Brown to Yellow | |
C6H14BaO2 | |
MFCD00050486 | |
barium isopropoxide, barium 2+ ; propan-2-olate, barium diisopropoxide, acmc-20alr0, barium ii isopropoxide, barium 2+ bis propan-2-olate, barium 2+ ion bis propan-2-olate | |
CC(C)[O-].CC(C)[O-].[Ba+2] | |
255.503 | |
255.52 | |
Barium isopropoxide |
24363-37-9 | |
Powder | |
Ba[OCH(CH3)2]2 | |
UN3205 | |
CPUJSIVIXCTVEI-UHFFFAOYSA-N | |
barium(2+);propan-2-olate | |
6101105 | |
Moisture Sensitive |
Safety and Handling
GHS H Statement
H314-H318-H228-H252-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Self-heating in large quantities
may catch fire.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310-P302+P352
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 4.2; Packing Group: III; Proper Shipping Name: ALKALINE EARTH METAL ALCOHOLATES, N.O.S.
RUO – Research Use Only