missing translation for 'onlineSavingsMsg'
Learn More

Barium peroxide, anhydrous, 84% min, Thermo Scientific Chemicals

41.00 € - 465.32 €

Chemical Identifiers

CAS 1304-29-6
Molecular Formula BaO2
Molecular Weight (g/mol) 169.325
MDL Number MFCD00003454
InChI Key ZJRXSAYFZMGQFP-UHFFFAOYSA-N
Synonym barium peroxide, barium dioxide, barium binoxide, barium superoxide, barium oxide, per, bario perossido di, bariumperoxid german, bariumperoxyde dutch, dioxyde de baryum french, barium peroxide ba o2
PubChem CID 14773
IUPAC Name barium(2+);peroxide
SMILES [O-][O-].[Ba+2]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11321539
View Documents
Thermo Scientific Alfa Aesar
014008.18
50 g
41.00 €
50g
Please to purchase this item. Need a web account? Register with us today!
11331539
View Documents
Thermo Scientific Alfa Aesar
014008.30
250 g
47.03 €
250g
Please to purchase this item. Need a web account? Register with us today!
11341539
View Documents
Thermo Scientific Alfa Aesar
014008.A1
1 kg
117.59 €
1kg
Please to purchase this item. Need a web account? Register with us today!
11351539
View Documents
Thermo Scientific Alfa Aesar
014008.A7
5 kg
465.32 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Barium peroxide is a specialty product for proteomics research. It is a strong oxidizing agent. It is also used for bleaching in a variety of commercial applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1304-29-6
169.325
ZJRXSAYFZMGQFP-UHFFFAOYSA-N
14773
[O-][O-].[Ba+2]
BaO2
MFCD00003454
barium peroxide, barium dioxide, barium binoxide, barium superoxide, barium oxide, per, bario perossido di, bariumperoxid german, bariumperoxyde dutch, dioxyde de baryum french, barium peroxide ba o2
barium(2+);peroxide

Specifications

450°C (decomposition)
White
84%
MFCD00003454
Insoluble in water
[O-][O-].[Ba+2]
169.325
169.34
Moisture sensitive
Barium peroxide
1304-29-6
Powder
BaO2
barium peroxide, barium dioxide, barium binoxide, barium superoxide, barium oxide, per, bario perossido di, bariumperoxid german, bariumperoxyde dutch, dioxyde de baryum french, barium peroxide ba o2
ZJRXSAYFZMGQFP-UHFFFAOYSA-N
barium(2+);peroxide
14773
Odorless
4.96 g/cm3
Videos
Safety and Handling

Safety and Handling

Keep away from food, drink and animal feeding stuffs. Avoid contact with skin and eyes. Take off immediately all contaminated clothing.

P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P312-P330-P370+P378q-P501c

H272-H302+H332

missing translation for 'dotInformation' : DOT Class: 5.1, Packaging Group: II

missing translation for 'einecsNumber' : 215-128-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.