missing translation for 'onlineSavingsMsg'
Learn More

Barium titanate(IV), 99%, Thermo Scientific Chemicals

58.90 € - 192.96 €

Chemical Identifiers

CAS 12047-27-7
Molecular Formula BaO3Ti
Molecular Weight (g/mol) 233.19
MDL Number MFCD00003447
InChI Key WNKMTAQXMLAYHX-UHFFFAOYSA-N
IUPAC Name barium(2+) oxotitaniumbis(olate)
SMILES [Ba++].[O-][Ti]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10694892
View Documents Promotion Details
Thermo Scientific Acros
196865000
500 g Plastic bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10226400
View Documents Promotion Details
Thermo Scientific Acros
196860025
2.5 kg Plastic bottle
192.96 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12047-27-7
233.19
WNKMTAQXMLAYHX-UHFFFAOYSA-N
[Ba++].[O-][Ti]([O-])=O
BaO3Ti
MFCD00003447
barium(2+) oxotitaniumbis(olate)

Specifications

1625.0°C
White
100.0
0.988 to 1.000 (BaO+ SrO)/TiO2 Mole Ratio
BaTiO3
15, 996
WNKMTAQXMLAYHX-UHFFFAOYSA-N
barium(2+) oxotitaniumbis(olate)
233.24
0.5% max.
6.0800g/mL
12047-27-7
98.5
Powder
BaO3Ti
MFCD00003447
Solubility in water: insoluble. Other solubilities: insoluble in alkalies, slightly soluble in diluted acids, soluble in concentrated h2so4 and hf
[Ba++].[O-][Ti]([O-])=O
233.19
99%
6.08
Barium titanate(IV), 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 234-975-0

missing translation for 'rtecsNumber' : XR1437333

missing translation for 'tsca' : TSCA

Product Identifier
  • Barium titanate(IV)

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only