missing translation for 'onlineSavingsMsg'
Learn More

Barium titanium oxide, 99%, Thermo Scientific Chemicals

Ferroelectric ceramics, single crystals, storage devices, dielectric amplifiers

35.58 € - 192.96 €

Chemical Identifiers

CAS 12047-27-7
Molecular Formula BaO3Ti
Molecular Weight (g/mol) 233.19
MDL Number MFCD00003447
InChI Key WNKMTAQXMLAYHX-UHFFFAOYSA-N
Synonym Barium titanate
IUPAC Name barium(2+) oxotitaniumbis(olate)
SMILES [Ba++].[O-][Ti]([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11331719
View Documents
Thermo Scientific Alfa Aesar
012348.18
50 g
35.58 €
50g
Please to purchase this item. Need a web account? Register with us today!
11351719
View Documents
Thermo Scientific Alfa Aesar
012348.36
500 g
63.82 €
500g
Please to purchase this item. Need a web account? Register with us today!
11341719
View Documents
Thermo Scientific Alfa Aesar
012348.A3
2 kg
192.96 €
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Barium titanium oxide is a dielectric ceramic used for capacitors. It is a piezoelectric material for microphones and other transducers. Finds applications in nonlinear optics and photorefractive applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12047-27-7
233.19
WNKMTAQXMLAYHX-UHFFFAOYSA-N
barium(2+) oxotitaniumbis(olate)
BaO3Ti
MFCD00003447
Barium titanate
[Ba++].[O-][Ti]([O-])=O

Specifications

1620°C
Powder
MFCD00003447
Barium titanate
WNKMTAQXMLAYHX-UHFFFAOYSA-N
barium(2+) oxotitaniumbis(olate)
233.19
Odorless
(metals basis)
12047-27-7
BaO3Ti
14,1000
Insoluble in water, alkalies; Slightly soluble in dilute acids; Soluble in concentrated acids
[Ba++].[O-][Ti]([O-])=O
233.19
99%
5.85 g/mL
Barium titanium oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

missing translation for 'einecsNumber' : 234-975-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.