missing translation for 'onlineSavingsMsg'
Learn More

Barium titanium oxide, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

60.30 € - 429.14 €

Chemical Identifiers

CAS 12047-27-7
Molecular Formula BaO3Ti
Molecular Weight (g/mol) 233.19
MDL Number MFCD00003447
InChI Key WNKMTAQXMLAYHX-UHFFFAOYSA-N
Synonym Barium titanate
IUPAC Name barium(2+) oxotitaniumbis(olate)
SMILES [Ba++].[O-][Ti]([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11320869
View Documents
Thermo Scientific Alfa Aesar
010652.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15405965
View Documents
Thermo Scientific Alfa Aesar
010652.14
25 g
124.62 €
25g
Please to purchase this item. Need a web account? Register with us today!
11310869
View Documents
Thermo Scientific Alfa Aesar
010652.22
100 g
429.14 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12047-27-7
233.19
WNKMTAQXMLAYHX-UHFFFAOYSA-N
barium(2+) oxotitaniumbis(olate)
BaO3Ti
MFCD00003447
Barium titanate
[Ba++].[O-][Ti]([O-])=O

Specifications

1620°C
White
BaO3Ti
14,1000
WNKMTAQXMLAYHX-UHFFFAOYSA-N
barium(2+) oxotitaniumbis(olate)
233.19
Puratronic™
5.85g/mL
Barium titanium oxide
12047-27-7
Powder
MFCD00003447
Barium titanate
[Ba++].[O-][Ti]([O-])=O
233.19
99.99%
Odorless
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'rtecsNumber' : XR1437333

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.