Learn More
Thermo Scientific™ Barnstead™ Hose Nipple Cartridges
Purify water up to 75 L/hr using these hose nipple cartridges, ideal for routine lab work including pretreatment.
Brand:  Thermo Scientific™ D8908
 View more versions of this product
Product Code. 15392178
  • 237.00 € / 1 unit
Estimated Shipment: 07-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Purify water volumes up to 75L/hr using cost-efficient Thermo Scientific™ Barnstead™ Hose Nipple Cartridges. Ideal for softening, deionization, and organic and chlorine removal applications, these cartridges are for use with the simple cartridge holder that easily mounts to the wall. Designed for non-pressure applications, these cartridges include 3/8 in. barb hose connections on each end. They provide the ability to see when resin is exhausted in specific cartridges by a color change indicator. Constructed of 100% virgin polypropylene cartridge, the system is customizable and an ideal option for routine laboratory work.
  • Ideal for small volume applications
  • Same superior quality resins used in B-Pure cartridges
  • Constructed of 100% virgin polypropylene cartridge
  • Includes 3/8″ barb hose connections on each end D8822, D8950, and D8951 cartridges include tapered straight nipple for 3/8″ ID tubing
  • Color change indicator that identifies when resin is exhausted in specific cartridges
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Full Size Macroreticular, Cation Removal
Cartridge, Hose Nipple
Macroreticular Cation Exchange
Barnstead Water Purification System
Up to 75 L/hr.
Macroreticular Cation Exchange
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Barnstead™ Hose Nipple Cartridges >