Learn More
Base Oil 75, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep BASE75
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- Base Oil 75 Blank
- 500 mL Volume
- Certified Base Oil
- Organometallic Oil Standard
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Specifications
Base Oil 75 Blank | |
500 mL | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Base Oil 75 |
Triple-leached LDPE bottle | |
With Certificate of Analysis | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Product Content Correction
Product Title
Product intended for laboratory use only
Spot an opportunity for improvement?Share a Content Correction