Learn More
Base Oil 75, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
45.45 € - 147.00 €
Specifications
Chemical Name or Material | Base Oil 75 Blank |
---|---|
Packaging | Triple-leached LDPE bottle |
Certificate of Analysis | With Certificate of Analysis |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15668235
|
SPEX Certiprep
BASE75 |
500 mL | Triple-leached LDPE bottle |
45.45 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11450259
|
SPEX Certiprep
BASE75-G |
1 gal. | Triple-leached LDPE bottle | N/A | |||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- Base Oil 75 Blank
- Certified Base Oil
- Organometallic Oil Standard
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Specifications
Base Oil 75 Blank | |
With Certificate of Analysis | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Triple-leached LDPE bottle | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Base Oil 75 |
Safety and Handling
Causes skin irritation, serious eye irritation
missing translation for 'dotInformation' : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.