missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ White Glove Delivery per unit

Delivery on the ground floor or on a different floor with freight elevator inkl. unpacking and taking back of packaging. Shipment with stair climb to be ordered when needed.

Brand:  Eppendorf™ 2277000002

 View more versions of this product

Product Code. 15773685

  • 653.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • White glove delivery per unit
  • Delivery on the ground floor or on a different floor with freight elevator inkl. unpacking and taking back of packaging
  • Shipment with stair climb to be ordered when needed
TRUSTED_SUSTAINABILITY
Specifications

Specifications

White glove delivery per unit (>180 kg)
White glove delivery (shipment with stair climb to be added when needed)
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Eppendorf™ White Glove Delivery per unit > <100 kg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.