Learn More
Thermo Scientific™ Pierce™ 3x DYKDDDDK Peptide
Tricosa-peptide for competitively eluting DYKDDDK-tagged fusion proteins from immobilized supports
Brand: Thermo Scientific™ A36805
292.68 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Pierce 3X DYKDDDDK Peptide is a lyophilized peptide with the sequence Met-Asp-Tyr-Lys-Asp-His-Asp-Gly-Asp-Tyr-Lys-Asp-His-Asp-Ile-Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys (MDYKDHDGDYKDHDIDYKDDDDK). It has a molecular weight of 2861.9 g/mol. For use, reconstitute to 1.5 mg/mL in PBS.
This tricosa-peptide can be used to competitively elute DYKDDDDK-tagged fusion proteins from immobilized anti-DYKDDDDK affinity supports, especially when a DYKDDDDK-tagged protein is sensitive to low pH. At a 1.5 mg/mL concentration, complete release of tagged protein can be achieved in 5–10 minutes at ambient temperature.
Note: Pierce 3X DYKDDDDK Peptide may interfere with protein determination assays and absorbance at 280 nm. However, the peptide can be removed from the eluate by desalting (e.g., with Zeba Spin Desalting Columns, Cat. No. 89891).

Specifications
Pierce™ | |
Recombinant | |
5 mg lyophilized peptide. Store at -5 to -30°C. | |
Wet Ice | |
Lyophilized Powder |
Peptides | |
5 mg | |
Lyophilized Powder | |
Protein Purification and Isolation | |
Affinity Resin |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
AB4603961 | Certificate of Analysis | 31/01/2025 | A36805 |
AA4559771 | Certificate of Analysis | 16/12/2024 | A36805 |
ZK4509201 | Certificate of Analysis | 26/10/2024 | A36805 |
ZJ4476321 | Certificate of Analysis | 15/10/2024 | A36805 |
ZH4390361 | Certificate of Analysis | 08/08/2024 | A36805 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.