Learn More
Cleaver Scientific Basic Gel Documentation Hood
A low cost, simple to use gel documentation hood for use with a mobile phone. Reduces background from ambient light when using blue light transilluminators.
Brand:  Cleaver Scientific GDH-BASIC
Product Code. 16684193
  • 129.00 € / Each
Estimated Shipment: 22-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Place the hood over a Blue light Transilluminator
  • Use a phone to zoom in and take an image
  • Transfer the images from phone to analysis software
  • Ideal for education use or where low costs are important
Specifications
Specifications
Mobile Phone
GDH-BASIC
Gel Documentation Hood
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cleaver Scientific Basic Gel Documentation Hood > GDH-BASIC