Learn More
Bathophenanthroline, 98+%, Thermo Scientific Chemicals
Description
Bathophenanthroline is widely used for determination of iron in serum and urine by colorimtetry. It acts as buffer layer to improve the efficiency of organic photovoltaic cells. It is very useful as a scavenger reagent, for the removal of traces of iron and copper from reagent solutions, which is used in the trace metal determination. Iron bathophenanthroline complex is involved as a redox mediating agent for imaging surface immobilized deoxyribonucleic acid (DNA) by using Scanning Electrochemical Microscopy (SECM).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Bathophenanthroline is widely used for determination of iron in serum and urine by colorimtetry. It acts as buffer layer to improve the efficiency of organic photovoltaic cells. It is very useful as a scavenger reagent, for the removal of traces of iron and copper from reagent solutions, which is used in the trace metal determination. Iron bathophenanthroline complex is involved as a redox mediating agent for imaging surface immobilized deoxyribonucleic acid (DNA) by using Scanning Electrochemical Microscopy (SECM).
Solubility
Soluble in organic solvents. Slightly soluble in acidic aqueous solutions. Insoluble in neutral or alkaline solutions.
Notes
Store in cool place. Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 218°C to 222°C |
Quantity | 1 g |
Beilstein | 261048 |
Solubility Information | Soluble in organic solvents. Slightly soluble in acidic aqueous solutions. Insoluble in neutral or alkaline solutions. |
Formula Weight | 332.41 |
Percent Purity | ≥98% |
Chemical Name or Material | Bathophenanthroline |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
T26I001 | Certificate of Analysis | 02/08/2022 | A14258.03,A14258.14,A14258.06 |
10241738 | Certificate of Analysis | 11/07/2022 | A14258.03,A14258.14,A14258.06 |
10238799 | Certificate of Analysis | 12/04/2022 | A14258.03,A14258.14,A14258.06 |
N09I007 | Certificate of Analysis | 12/04/2022 | A14258.03,A14258.14,A14258.06 |
Z14D007 | Certificate of Analysis | 22/11/2021 | A14258.03,A14258.14,A14258.06 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.