Learn More
Tocris Bioscience™ Bax channel blocker
Allosteric inhibitor of Bax channel activation
Brand:  Tocris Bioscience™ 2160/10
 View more versions of this product
Product Code. 18064642
  • 263.00 € / 10mg
Explore more special offers
Description
Description
Bax channel blocker is a allosteric inhibitor of Bax channel activation. Binds inactive Bax at allosteric site and inhibits tBID-mediated Bax activation (IC50 = 3.3 μM). Selectively inhibits Bax-dependent apoptosis. Potent inhibitor of Bax-mediated mitochondrial cytochrome c release (IC50 = 0.52 μM).
Specifications
Specifications
>98%
Bax channel blocker
Bax channel blocker
10 mg
540.12