missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Bax channel blocker
Allosteric inhibitor of Bax channel activation
263.00 € - 1050.00 €
Specifications
Purity | >98% |
---|---|
Product Type | Bax channel blocker |
Molecular Weight (g/mol) | 540.12 |
Inhibitors | Bax channel blocker |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18064642
|
Tocris Bioscience™
2160/10 |
10 mg |
263.00 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18074972
|
Tocris Bioscience™
2160/50 |
50 mg |
1050.00 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent inhibitor of Bax-mediated mitochondrial cytochrome c release (IC50 = 0.52 μM). Suggested to inhibit Bax channel formation/activity.Specifications
>98% | |
Bax channel blocker | |
540.12 | |
Bax channel blocker |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title