Learn More
Berkshire™ BCR™ Polyester Full-Finger Cleanroom Glove Liners
BIOPHARMA
Continuous filament knitted polyester construction is thin yet durable, while the full-finger design provides total protection for those with severe glove sensitivities.
Brand:  Berkshire™ BGL3.20L
32.02 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 12950975
  • 34.55 € / 10 pairs
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Reduces perspiration and contact dermatitis commonly associated with wearing latex, urethane, PVC, or nitrile gloves
  • Full-finger design provides total protection for those with severe glove sensitivities
  • Can be worn alone as inspection gloves to prevent fingerprints on critical surfaces
  • 100% continuous filament knitted polyester is thin, yet durable
  • Ambidextrous design can be used on either hand
  • Gloves can be repeatedly laundered for reuse
  • Suitable for use in ISO Class Cleanrooms
Specifications
Specifications
Uncoated
Reusable
Polyester
Full
Blue
BGL3
White
Ambidextrous
Large
Laboratories and Manufacturing Environments
Knit