Learn More
BD Bacto™ Dehydrated Agar
Purified solidifying agent. BD Difco™ Bacto™ Dehydrated Agar is purified to remove extraneous matter.
Brand:  BD 214050
 View more versions of this product
Product Code. 11768223
  • 111.00 € / 100g
Edge
Explore more special offers
Description
Description
Purified solidifying agent. BD Difco™ Bacto™ Dehydrated Agar is purified to remove extraneous matter.
  • Extraneous matter, pigmented portions and salts are reduced to minimum
  • Fine, light colored granules
  • Convenient for weighing and handling
Determines motility and growth of anaerobes and microaerophiles
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO/DIS, 11014
Agar
Determination of motility and the growth of anaerobes and microaerophiles
100 g
1.5%
Dehydrated
Bottle
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD Bacto™ Dehydrated Agar > Dehydrated Agar; 100g