missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Rabbit Coagulase Plasma

Qualitative determination of Staphylococci pathogenicity using direct tube method

324.00 €

Specifications

Species Rabbit
Certifications/Compliance CE
Content And Storage 2°C to 8°C
Form Lyophilized
Product Type Coagulase Rabbit Plasma
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11758852
View Documents
BD
240826
10 x 15 mL
324.00 €
150mL
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
 
Description

Description

Coagulase binds plasma fibrinogen, causing the organisms to agglutinate or plasma to clot. BBL Coagulase Plasma, Rabbit and BBL Coagulase Plasma Rabbit with EDTA are recommended for performing the direct tube test. At the time of use, test both positive and negative control cultures to check performance of the coagulase plasma.

  • Product type: diagnostic reagents and adjuvants
  • Groupings: C Products
  • Type: Coagulase Testing
  • Shelf Life: 1095 days
  • Label has Natural Rubber Latex statement

Certifications

CE marked

Specifications

Specifications

Rabbit
2°C to 8°C
Coagulase Rabbit Plasma
CE
Lyophilized
Qualitative determination of Staphylococci pathogenicity using direct tube method
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 1095 Days

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.