Learn More
BD BBL™ Rabbit Coagulase Plasma
Qualitative determination of Staphylococci pathogenicity using direct tube method
Brand: BD 240826
Description
Coagulase binds plasma fibrinogen, causing the organisms to agglutinate or plasma to clot. BBL Coagulase Plasma, Rabbit and BBL Coagulase Plasma Rabbit with EDTA are recommended for performing the direct tube test. At the time of use, test both positive and negative control cultures to check performance of the coagulase plasma.
- Product type: diagnostic reagents and adjuvants
- Groupings: C Products
- Type: Coagulase Testing
- Shelf Life: 1095 days
- Label has Natural Rubber Latex statement
Certifications
CE marked

Specifications
Rabbit | |
2°C to 8°C | |
Coagulase Rabbit Plasma | |
10 x 15 mL |
CE | |
Lyophilized | |
Qualitative determination of Staphylococci pathogenicity using direct tube method |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.