Learn More
BD Difco™ Microbial Content Test Agar
Recommended for the detection and enumeration of microorganisms present on surfaces of sanitary importance
Brand:  BD 255320
 View more versions of this product
Product Code. 11768982
  • 230.00 € / 500g
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Certifications
CCAM
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CCAM
Dehydrated
Detection and enumeration of microorganisms
500 g
2°C to 8°C
Microbial Content Test Agar
Bottle