missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Microbial Content Test Agar
Recommended for the detection and enumeration of microorganisms present on surfaces of sanitary importance
230.00 € - 417.00 €
Specifications
Certifications/Compliance | CCAM |
---|---|
Form | Dehydrated |
Media Type | Microbial Content Test Agar |
For Use With (Application) | Detection and enumeration of microorganisms |
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
11768982
|
BD
255320 |
Bottle | 500 g |
230.00 €
500g |
Estimated Shipment: 07-11-2024
Log in to see stock. |
||||
11758982
|
BD
255310 |
Pail | 2 kg |
417.00 €
2kg |
Estimated Shipment: 07-11-2024
Log in to see stock. |
||||
Description
Certifications
CCAM
Specifications
CCAM | |
Dehydrated | |
Microbial Content Test Agar | |
Detection and enumeration of microorganisms |
Safety and Handling
missing translation for 'shelfLife' : 1460 Days