Learn More
BD Difco™ Neisseria Meningitidis Antisera
For serotyping Neisseria meningitidis in slide agglutination tests
Brand:  BD 222311
 View more versions of this product
Product Code. 11758333
  • 209.00 € / 1mL
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Description
Description
  • Stable, lyophilized, polyclonal rabbit antisera
  • Contains 0.1% sodium azide as preservative
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For in vitro diagnostic use
Neisseria meningitidis Antiserum Group D
Antiserum
1 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For in vitro diagnostic use.