missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Neisseria Meningitidis Antisera
For serotyping Neisseria meningitidis in slide agglutination tests
Brand: BD 222521
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Description
- Stable, lyophilized, polyclonal rabbit antisera
- Contains 0.1% sodium azide as preservative
Specifications
For in vitro diagnostic use | |
Antiserum | |
1 mL |
N. Meningitidis Antiserum Group Z prime (Z' ) | |
Bottle |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- BD Difco Neisseria Meningitidis Antiserum Group Z', Bottle, 1 ml
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium azide
For in vitro diagnostic use.