missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Neisseria Meningitidis Antisera
For serotyping Neisseria meningitidis in slide agglutination tests
Brand: BD 229101
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Description
- Stable, lyophilized, polyclonal rabbit antisera
- Contains 0.1% sodium azide as preservative
Specifications
For in vitro diagnostic use | |
Neisseria meningitidis Antiserum Poly 2, Groups X, Y, Z | |
Antiserum | |
3 mL |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P270-Do not eat, drink or smoke when using this product.
- P264-Wash thoroughly after handling.
- P330-Rinse mouth.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : sodium azide
For in vitro diagnostic use.