Learn More
BD Disposable Syringes with Luer-Lok™ Tips
Unique integrated BD Luer-Lok™ tip syringe with clear barrel. BD Medical™ Disposable Syringes with Luer-Lok™ Tips are sterile and disposable.
Brand: BD 309628
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Unique integrated BD Luer-Lok™ tip syringe with clear barrel. BD Medical™ Disposable Syringes with Luer-Lok™ Tips are sterile and disposable.
- Sterile syringe for single use
- Without needle
- Clear barrel (polycarbonate or polypropylene) with bold scale markings
- Tapered plunger rod for ease of aspiration
- Positive plunger rod stop
- Added BD Luer-Lok™ thread for increased secure connection
- DNR (Latex) free
- Shelf life: 1825 days
Specifica
Syringe | |
No Needle Included | |
Luer Lock |
Single-use | |
1 mL | |
Sterile |