Learn More
BD DrySlide™ Reagent Slides
A convenient method for bacterial biochemical analysis. Self-contained, ready-to-use test slides consist of coated, 35mm film or impregnated paper, which operates as a carrier matrix for microbiological testing reagents.
Brand:  BD 231749
 View more versions of this product
Product Code. 11722994
  • 860.00 € / Pack of 25
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • One-step procedure —no need for rehydrating
  • No thin paper strips to manipulate and no glass slides are involved
  • Each individual slide contains four reaction areas
  • DrySlide Indole is a 30-second, spot indole test that uses DMABA (5% p-dimethylaminobenzaldehyde) formulation to test for indole-producing bacteria
  • DrySlide Nitrocefin is a qualitative test to detect the presence of bacteria that produces β-lactamase and meets NCCLS guidelines; each slide contains a reagent cassette impregnated with nitrocefin
  • DrySlide Oxidase with each slide is film-coated with an oxidase reagent (N, N, N',N'-tetramethyl-p-phenylenediamine dihydrochloride) to test for the presence of cytochrome C oxidase activity in bacteria; a positive result produces a purple color within 20 seconds
  • DrySlide PYR Kit has L-pyrrolidonyl-β-naphthylamide that is used for the rapid presumptive identification of Group D Enterococci and Strep Group A; includes 15 slides and 15, 0.5mL crush ampules
Order Info
Slides are individually wrapped and supplied in convenient dispenser packs
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nitrocefin Reagent
CE
15° to 30°C
25 Pack
Used for detecting beta-Lactamase production by bacteria