Learn More
BD DrySlide™ Reagent Slides
A convenient method for bacterial biochemical analysis. Self-contained, ready-to-use test slides consist of coated, 35mm film or impregnated paper, which operates as a carrier matrix for microbiological testing reagents.
735.00 € - 1325.00 €
Product Code | Brand | Quantity | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Type | Price | Quantity & Availability | |||||
11792984
|
BD
231746 |
75 Pack | Oxidase Reagent |
750.00 €
Pack of 75 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11702994
|
BD
231747 |
15 Pack | PYR Kit |
1325.00 €
Pack of 15 |
Estimated Shipment: 22-05-2025
Log in to see stock. |
|||||
11712994
|
BD
231748 |
75 Pack | Indole Reagent |
735.00 €
Pack of 75 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11722994
|
BD
231749 |
25 Pack | Nitrocefin Reagent |
860.00 €
Pack of 25 |
Estimated Shipment: 22-05-2025
Log in to see stock. |
|||||
Description
- One-step procedure —no need for rehydrating
- No thin paper strips to manipulate and no glass slides are involved
- Each individual slide contains four reaction areas
- DrySlide Indole is a 30-second, spot indole test that uses DMABA (5% p-dimethylaminobenzaldehyde) formulation to test for indole-producing bacteria
- DrySlide Nitrocefin is a qualitative test to detect the presence of bacteria that produces β-lactamase and meets NCCLS guidelines; each slide contains a reagent cassette impregnated with nitrocefin
- DrySlide Oxidase with each slide is film-coated with an oxidase reagent (N, N, N',N'-tetramethyl-p-phenylenediamine dihydrochloride) to test for the presence of cytochrome C oxidase activity in bacteria; a positive result produces a purple color within 20 seconds
- DrySlide PYR Kit has L-pyrrolidonyl-β-naphthylamide that is used for the rapid presumptive identification of Group D Enterococci and Strep Group A; includes 15 slides and 15, 0.5mL crush ampules
Order Info
Slides are individually wrapped and supplied in convenient dispenser packs
Specifications
CE |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BD DrySlide™ Reagent Slides