missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Horizon™ BV605 Annexin V
A sensitive probe for identifying apoptotic cells, binding to negatively charged phospholipid surfaces with a higher affinity for phosphatidylserine (PS) than most other phospholipids
Brand: BD 563974
415.16 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description

Specifications
Flow Cytometry | |
BV605 | |
Buffered Solution |
100 Tests | |
Store undiluted at 4°C |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Horizon™ BV605 Annexin V > 100 Tests
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction