Learn More
BD Perm/Wash™ Buffer
Can be used in intracellular cytokine staining to permeabilize cells and as an antibody diluent and cell wash buffer
Brand: BD 554723
247.17 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- 10X stock solution containing both Fetal Bovine Serum (FBS) and saponin
It is not uncommon for the color of this product to vary from lot to lot. A small amount of insoluble precipitate is also common. Color variation and/or precipitate do not affect product performance.
Specifications
Fetal Bovine Serum (FBS) Solution 10X and Saponin | |
7.2 to 7.6 | |
Perm or Wash Buffer | |
Liquid |
Can be used in intracellular Cyto kine staining to permeabilize cells and as an Antibody diluent and cell Wash Buffer | |
4°C | |
100 mL |