Learn More
Heidolph Instruments Stirring Bar Sets
Stirring bars available in oval and cylindrical shape sets
Brand:  Heidolph Instruments 509-56000-00
 View more versions of this product
Product Code. 10157841
  • 26.00 € / Each
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Oval Stir Bars:
  • Shape is similar to that of the flask and ensures complete mixing
  • Equipped with an oval magnetic core, more force is available than with a cylindrical magnetic core
  • For applications in:
    • Round bottom flasks e.g. for reaction mixtures
Cylindrical Stir Bars:
  • Smooth operating conditions at optimum contact area
  • Long lifetime
  • For applications in:
    • Beakers
    • Crystallizing dishes
    • Heating bathes
    • Other flat bottom vessels
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
25, 40, 50 mm
25, 40, 50 mm
1/Pk.
White
PTFE
PTFE
Cylinder
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Heidolph Instruments Stirring Bar Sets > Size: 25, 40, 50 mm; Cylindrical