missing translation for 'onlineSavingsMsg'
Learn More

BD Pharmingen™ IHC Zinc Fixative

An alternative to formalin fixation of tissue for paraffin embedded sections

Brand:  BD 552658


 View more versions of this product

Product Code. 15886838

  • 381.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Ten percent neutral buffered formalin fixes tissue by cross-linking proteins, potentially causing a conformational change or “masking” of the antigenic epitope and leading to reduced or absent antibody reactivity. Different antigen retrieval techniques are used to unmask the epitope; however, many times it is not possible to retrieve the epitope and get optimal immunohistochemical staining. In these situations the use of IHC Zinc fixative, which is a milder fixative than formalin, is recommended.

  • Zinc-fixed paraffin embedded sections show very good immunostaining with many leukocytes markers and depict tissue morphology comparable to formalin-fixation
  • Use of antigen retrieval techniques on the zinc-fixed paraffin sections may further enhance the antibody reactivity in some cases

There are a few antigens that are not preserved by zinc-fixation. In these situations use of fresh frozen tissue for immunohistochemical staining is the only alternative.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

100°C
Room temperature
Fixative
Immunohistochemical staining
Liquid
Clear
10 X
500mL
7.2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 10X Zinc Fixative (Formalin Free)

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : zinc acetate, zinc chloride

SDS
Documents

Documents

Certificates
Special Offers

Special Offers