Learn More
BD Pharmingen™ IHC Zinc Fixative
An alternative to formalin fixation of tissue for paraffin embedded sections
159.00 € - 381.00 €
Specifications
Color | Clear |
---|---|
Content And Storage | Room temperature |
Product Type | Fixative |
For Use With (Application) | Immunohistochemical staining |
Form | Liquid |
Product Code | Brand | Dilution | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dilution | Quantity | Price | Quantity & Availability | ||||
10400275
|
BD
550523 |
1 X | 1000mL |
159.00 €
Each |
Estimated Shipment: 02-12-2024
Log in to see stock. |
||||
15886838
|
BD
552658 |
10 X | 500mL |
381.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Ten percent neutral buffered formalin fixes tissue by cross-linking proteins, potentially causing a conformational change or “masking” of the antigenic epitope and leading to reduced or absent antibody reactivity. Different antigen retrieval techniques are used to unmask the epitope; however, many times it is not possible to retrieve the epitope and get optimal immunohistochemical staining. In these situations the use of IHC Zinc fixative, which is a milder fixative than formalin, is recommended.
- Zinc-fixed paraffin embedded sections show very good immunostaining with many leukocytes markers and depict tissue morphology comparable to formalin-fixation
- Use of antigen retrieval techniques on the zinc-fixed paraffin sections may further enhance the antibody reactivity in some cases
There are a few antigens that are not preserved by zinc-fixation. In these situations use of fresh frozen tissue for immunohistochemical staining is the only alternative.
Specifications
Clear | |
Fixative | |
Liquid |
Room temperature | |
Immunohistochemical staining |