Learn More
BD Recombinant Mouse TNF
Potent multifunctional cytokine containing 156 amino acid residues
Brand: BD 554589
580.65 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Glycosylated protein
- Exerts regulatory and cytotoxic effects on wide range of normal lymphoid and non-lymphoid cells and tumor cells
- Storage buffer: frozen aqueous buffered solution containing BSA
Bioassay, ELISA standard, blocking

Specifications
200μg/mL | |
Frozen aqueous Buffered solution containing BSA | |
10μg | |
Mouse |
Bioassay, Blocking Agent, ELISA Standard | |
TNF-alpha Recombinant Protein | |
-80°C | |
Recombinant |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For research use only.