missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Recombinant Rat Tumor Necrosis Factor (TNF-α)
Potent multifunctional cytokine
Brand: BD
369.46 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Molecular weight: 17.4kD protein
- Contains 157 amino acid residues
- Reactivity: rat
- Storage buffer: frozen aqueous buffered solution containing BSA
- Exerts regulatory and cytotoxic effects on normal lymphoid and non-lymphoid cells and tumor cells
Bioassay, ELISA standard

Specifications
100μg/mL | |
Frozen aqueous Buffered solution containing BSA | |
5μg | |
Rat |
Bioassay, ELISA Standard | |
TNF-alpha Recombinant Protein | |
-80°C | |
Recombinant |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Recombinant Rat Tumor Necrosis Factor (TNF-α) > Recombinant Rat Tumor Necrosis Factor (TNF-alpha); 100μg/mL; 5μg
For research use only.
Spot an opportunity for improvement?Share a Content Correction