missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Lab-Aire™ II Glassware Dryers Accessories

Replacement pegs for use with Lab-Aire II benchtop and wallmount dryer models.

Brand:  Bel-Art™ F18931-0001

 View more versions of this product

Product Code. 13213419

  • 34.35 € / Pack of 19
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Available as small (3 in.), medium (4 in.) and large (5 in.) pegs
  • Pegs can be moved and repositioned to hold variety of labware including beakers, flasks, bottles, test tubes and cylinders
Specifications

Specifications

Replacement Peg
Lab-Aire II benchtop and wallmount dryers
3 in.
Not Autoclavable
Small
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers